missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Acetate TS, 9.5% (w/v), Ricca Chemical
Meets USP Specifications
$186.87 - $186.87
Chemical Identifiers
| CAS | 6080-56-4 |
|---|---|
| Molecular Formula | C4H12O7Pb |
| Molecular Weight (g/mol) | 379.30 |
| MDL Number | MFCD00150023 |
| InChI Key | MCEUZMYFCCOOQO-UHFFFAOYSA-L |
| Synonym | lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate |
| PubChem CID | 22456 |
| ChEBI | CHEBI:33112 |
| IUPAC Name | λ2-lead(2+) diacetate trihydrate |
| SMILES | O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
Chemical Identifiers
| 6080-56-4 | |
| 379.30 | |
| MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
| 22456 | |
| λ2-lead(2+) diacetate trihydrate |
| C4H12O7Pb | |
| MFCD00150023 | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| CHEBI:33112 | |
| O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
Specifications
| Colorless | |
| 89.0,8.51 | |
| C4H12O7Pb | |
| lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
| O.O.O.[Pb++].CC([O-])=O.CC([O-])=O | |
| 379.30 | |
| CHEBI:33112 | |
| Laboratory | |
| Aqueous Solution | |
| 120 mL | |
| 9.5% (w/v) |