CAS RN 6080-56-4
Lead(II) acetate trihydrate, ACS, 99.0-103.0%
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis)
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 IUPAC Name: lead(2+);diacetate;trihydrate SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead(II) acetate trihydrate, 99+%, for analysis
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead(II) acetate trihydrate, 99%
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 IUPAC Name: lead(2+);diacetate;trihydrate SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead(II) acetate trihydrate, 99+%, ACS reagent
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead Acetate, 10% (w/v) with 1% (v/v) Acetic Acid, Ricca Chemical
Chemical Name or Material: Lead Acetate CAS: 6080-56-4 CAS Min %: 8.83 CAS Max %: 9.19 CAS: 7732-18-5 CAS Min %: 88.0 CAS Max %: 92.0 CAS: 64-19-7 CAS Min %: 0.93 CAS Max %: 0.96
Lead Acetate TS, 9.5% (w/v), Ricca Chemical
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.333 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 IUPAC Name: lead(2+);diacetate;trihydrate SMILES: CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]
Lead Acetate Trihydrate, ACS Reagent Grade, Ricca Chemical
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.333 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 IUPAC Name: lead(2+);diacetate;trihydrate SMILES: CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]
Lead Acetate, 5% (w/v), Ricca Chemical
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 IUPAC Name: λ2-lead(2+) diacetate trihydrate SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead Acetate, 2% (w/v), Ricca Chemical
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 IUPAC Name: λ2-lead(2+) diacetate trihydrate SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead Acetate, Trihydrate, ACS Grade, 99.0 to 103.0%, LabChem™
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 IUPAC Name: λ²-lead(2+) diacetate trihydrate SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Lead Acetate, For Arsenic, 10% (w/v), LabChem™
CAS: 6080-56-4 Molecular Formula: C4H12O7Pb Molecular Weight (g/mol): 379.30 MDL Number: MFCD00150023 InChI Key: MCEUZMYFCCOOQO-UHFFFAOYSA-L Synonym: lead acetate trihydrate,lead diacetate trihydrate,bleiazetat german,bis acetato trihydroxytrilead,unii-rx077p88ry,ccris 9031,lead acetate ii , trihydrate,lead acetate jan,acetic acid, lead +2 salt trihydrate,acetic acid, lead 2+ salt, trihydrate PubChem CID: 22456 ChEBI: CHEBI:33112 IUPAC Name: λ²-lead(2+) diacetate trihydrate SMILES: O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
- 1
- 2