missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate TS, 9.5% (w/v), Ricca Chemical

Catalog No. 42934
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
42934 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42934 Supplier Ricca Chemical Company Supplier No. RI42934

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meets USP Specifications

Chemical Identifiers

CAS 6080-56-4 , 7732-18-5
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless
CAS Min % 89.0, 8.51
CAS Max % 93.0, 8.85
Percent Purity 9.5%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 5.5 to 6.5
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 9.5% (w/v)
Chemical Name or Material Lead Acetate, Test Solution
Show More Show Less