missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Acetate, 5% (w/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 4.66, 93.0, 0.1 |
| CAS Max % | 4.86, 97.0, 0.1 |
| Percent Purity | 5% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Quantity | 1 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 5% (w/v) |
| Show More |