missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate, 2% (w/v), Ricca Chemical

Catalog No. 429132
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
429132 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 429132 Supplier Ricca Chemical Company Supplier No. 429132

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6080-56-4 , 7732-18-5 , 64-19-7
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color Colorless
CAS Min % 1.92, 96.0, 0.1
CAS Max % 2.0, 100.0, 0.1
Percent Purity 2%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 2% (w/v)
Chemical Name or Material Lead Acetate
Show More Show Less