Learn More
PIG™ Latching Drum Lid with Fast-Latch Ring
Latching drum lid to keep drums and hazardous waste storage containers securely closed and locked.
Supplier:  PIG™ DRM1072BK
 Afficher plus de versions de ce produit
Numéro de catalogue. 18366497
Il en reste null
Description
Description
  • For 55 gal. Steel Drums
  • Fast Latch
  • Fast-latch ring for easy installation
  • Adjustment rod to expand ring to fit reconditioned drums with larger diameters
  • Latching handle easily opens and closes with one hand
  • Nitrile gasket and locking mechanism seal lid to help reduce vapor emissions and keep drum contents dry and secure
  • Easy-access lid closes securely to help you meet hazardous waste regulations
  • Hinge stops keep funnel lid upright when open
  • Durable, chemical-resistant powder coat helps ensure long service life
  • Lockable latch helps prevent unauthorized access
  • Hinge pin is made for easy lid removal without loosening band around drum
  • Multiple lid color options let you segregate drums for safe, compliant filling
Spécifications
Identifiants chimiques
Latching Drum Lid
26.75 in.
11 cm
Non-sterile
68 cm
Spécifications
Latching Drum Lid
Non-sterile
68 cm
59 cm
11 cm
8.2 kg
Black
26.75 in.
23.25 in.
4.375 in.
18 lb.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS