missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIG™ Latching Drum Lid with Fast-Latch Ring
Latching drum lid to keep drums and hazardous waste storage containers securely closed and locked.
$1,149.50
Specifications
| Product Type | Latching Drum Lid |
|---|---|
| Sterility | Non-sterile |
| Length (English) | 26.75 in. |
| Length (Metric) | 68 cm |
| Height (Metric) | 11 cm |
Description
- For 55 gal. Steel Drums
- Fast Latch
- Fast-latch ring for easy installation
- Adjustment rod to expand ring to fit reconditioned drums with larger diameters
- Latching handle easily opens and closes with one hand
- Nitrile gasket and locking mechanism seal lid to help reduce vapor emissions and keep drum contents dry and secure
- Easy-access lid closes securely to help you meet hazardous waste regulations
- Hinge stops keep funnel lid upright when open
- Durable, chemical-resistant powder coat helps ensure long service life
- Lockable latch helps prevent unauthorized access
- Hinge pin is made for easy lid removal without loosening band around drum
- Multiple lid color options let you segregate drums for safe, compliant filling
Specifications
| Latching Drum Lid | |
| 26.75 in. | |
| 11 cm |
| Non-sterile | |
| 68 cm |