missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, Honeywell Fluka™

Numéro de catalogue. 6002820
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
6002820 1 L
6002821 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 6002820 Fournisseur Honeywell Chemicals Code fournisseur 697751L
Il en reste null

Tested according to Ph.Eur.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Identifiants chimiques

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Spécifications

Linear Formula CH3CH(OH)COOH
UN Number NONH for all modes of transport
Beilstein 1209341
Formula Weight 90.08g/mol
Density 1.209g/mL at 25°C
Quantity 1 L
Flash Point 113°C (235.4°F)
Chemical Name or Material Lactic acid
Hazard Statement P280-P305 + P351 + P338