missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ β-Lactamase, Bacillus cereus 569/H9

Catalog No. 426205500U
Change view
Click to view available options
Quantity:
500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
426205500U 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 426205500U Supplier MilliporeSigma™ Supplier No. 426205500U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful for assaying penicillins and cephalosporins

  • Native β-lactamase from Bacillus cereus 569/H9. Bacterial enzyme that destroys β-lactam antibiotics
  • R Phrase - R: 42/43
  • May cause sensitization by inhalation and skin contact
  • S Phrase - S: 36
  • Wear suitable protective clothing

Specifications

CAS 9073-60-3
Content And Storage -20°C
Form Lyophilized
Quantity 500 U
Synonym Penicillinase
Solubility Water
Product Type beta Lactamase