missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum, 14 gal.
GREENER_CHOICE

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

$95.68 - $104.06

Spécifications

Product Type Drum
Closure Included Yes
Capacity (English) 14 gal.
Capacity (Metric) 53.06 L
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Color Closure Type Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Color Closure Type Prix Quantity & Availability  
19338424
GREENER_CHOICE
Afficher les documents Détails de la promotion
Eagle™
1610BLK
Black Plastic Lever-lock
chaque for $95.68
Il en reste null
 
19338427
GREENER_CHOICE
Afficher les documents Détails de la promotion
Eagle™
1610MBLK
Black Metal Lever-lock
chaque for $104.06
Il en reste null
 
19338425
GREENER_CHOICE
Afficher les documents Détails de la promotion
Eagle™
1610M
Yellow Metal Lever-lock
chaque for $100.55
Il en reste null
 
Description

Description

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids
Spécifications

Spécifications

Drum
14 gal.
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
63.5 cm
Yes
53.06 L
HDPE
25 in.
10 lb.
Videos
FDS
Documents

Documents