missing translation for 'onlineSavingsMsg'
Learn More

L-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical

ME110, 2216-51-5, C10H20O

$288.24 - $22,176.97

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
IUPAC Name (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18603944
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $288.24
Only null left
 
18603949
View Documents Promotion Details
N/A 50 g Amber Glass Bottle
Each for $437.21
Only null left
 
18603945
View Documents Promotion Details
N/A 125 g Amber Glass Bottle
Each for $658.03
Only null left
 
18603946
View Documents Promotion Details
N/A 500 g Amber Glass Bottle
Each for $1,865.12
Only null left
 
18603947
View Documents Promotion Details
N/A 12 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18603948
View Documents Promotion Details
N/A 25 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical L-Menthol, Crystal, USP is used as a local anesthetic, a counterirritant and can help relieve minor throat irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

2216-51-5
156.27
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
C10H20O
NOOLISFMXDJSKH-KXUCPTDWSA-N
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

2216-51-5
41-44°C
-45° to -51°
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
156.27
USP
L-Menthol
100%
C10H20O
NOOLISFMXDJSKH-KXUCPTDWSA-N
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
98 to 102%
Crystalline Powder or Lumps
Videos
SDS
Documents

Documents