missing translation for 'onlineSavingsMsg'
Learn More

L-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18603945
Change view
Click to view available options
Quantity:
25 g
50 g
125 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603945 125 g Amber Glass Bottle
18603944 25 g Amber Glass Bottle
18603949 50 g Amber Glass Bottle
18603946 500 g Amber Glass Bottle
18603947 12 kg Amber Glass Bottle
18603948 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18603945 Supplier Spectrum Chemical Mfg Cor Supplier No. ME110125GM
Only null left

ME110, 2216-51-5, C10H20O

Spectrum™ Chemical L-Menthol, Crystal, USP is used as a local anesthetic, a counterirritant and can help relieve minor throat irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
IUPAC Name (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

CAS Max % 100%
Melting Range 41-44°C
Packaging Amber Glass Bottle
Quantity 125 g
Specific Rotation -45° to -51°
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material L-Menthol