missing translation for 'onlineSavingsMsg'
Learn More

L-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical

ME110, 2216-51-5, C10H20O

Supplier:  Spectrum Chemical Mfg Cor ME11012KGBL

Quantity 12 kg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18603947

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Spectrum™ Chemical L-Menthol, Crystal, USP is used as a local anesthetic, a counterirritant and can help relieve minor throat irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

2216-51-5
156.27
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
C10H20O
NOOLISFMXDJSKH-KXUCPTDWSA-N
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

Specifications

L-Menthol
100%
Amber Glass Bottle
12 kg
NOOLISFMXDJSKH-KXUCPTDWSA-N
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
98 to 102%
Crystalline Powder or Lumps
2216-51-5
41-44°C
C10H20O
-45° to -51°
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
156.27
USP
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions