missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ L.b. Cas12a Nuclease

Catalog No. A40001796
Change view
Click to view available options
Quantity:
200 pmol
2000 pmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40001796 200 pmol
A40001795 2000 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A40001796 Supplier Invitrogen™ Supplier No. A40001796

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L.b. Cas12a Nuclease for simple, quick and specific analysis of your samples.

L. b. (Lachnospiraceae bacterium) Cas12a Nuclease, also known as Cpf1, is an RNA-guided endonuclease derived from the Class II type V CRISPR-Cas system. It exhibits a high degree of specificity in targeting and cleaving particular DNA sequences in genome editing. Additionally, Cas12a nuclease possesses collateral cleavage activity, resulting in the non-specific cleavage of neighboring DNA upon target recognition.

The DNA cleavage capability of Cas12a nuclease has been leveraged for various applications, including genome editing, gene regulation, and nucleic acid detection. By programming Cas12a with specific guide RNAs, targeted modifications such as gene knockout, gene insertion, or point mutations can be introduced into the target DNA sequence.

Disclaimer: The use of this product for certain applications may require a license under third party intellectual property rights and it is buyer's sole responsibility to obtain any required license for such use. Thermo Fisher Scientific makes no representation or warranty that the use of this product is free from third party intellectual property rights, and will not indemnify the buyer against any third party infringement claim arising from buyer's use of the product.

Specifications

Concentration 10 μM
Quantity 200 pmol
Product Type Stand-alone enzyme
Research Category Nucleic Acid Analysis
Form Liquid

For Research Use Only.