Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
$65.07 - $178.39
Specifications
| Material | Glass |
|---|---|
| Product Type | Solid Pennyhead Stopper |
| For Use With (Equipment) | Flasks, mixing cylinders and separatory funnels |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
K850100-0019
|
DWK Life Sciences
8501000019 |
19 |
Each for $87.96
|
|
|||||
|
K850100-0022
|
DWK Life Sciences
8501000022 |
Each for $98.36
|
|
||||||
|
K850500-1018
|
DWK Life Sciences
8505001018 |
10/18 |
Each for $87.43
|
|
|||||
|
K850100-0027
|
DWK Life Sciences
8501000027 |
27 |
Each for $103.21
|
|
|||||
|
0254327
|
DWK Life Sciences
W8501000038 |
38 |
Each for $178.39
|
|
|||||
|
K850500-2425
|
DWK Life Sciences
8505002425 |
24/25 |
Each for $166.57
|
|
|||||
|
K850500-1922
|
DWK Life Sciences
8505001922 |
19/22 |
Each for $95.75
|
|
|||||
|
22-301346
|
DWK Life Sciences
8501000009 |
Case for $65.07
|
|
||||||
|
22-301347
|
DWK Life Sciences
8501000013 |
Case for $70.18
|
|
||||||
Description
- All stoppers are solid except Standard Taper stopper sizes 32 and 38, which are hollow
- Made in accordance with ASTM Standard E694
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
| Glass | |
| Solid Pennyhead Stopper | |
| Flasks, mixing cylinders and separatory funnels |