Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
Supplier:  DWK Life Sciences W8501000038
 View more versions of this product
Catalog No. 0254327
Only null left
Explore available promotions
Description
Description
  • All stoppers are solid except Standard Taper stopper sizes 32 and 38, which are hollow
  • Made in accordance with ASTM Standard E694
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Solid Pennyhead Stopper
38
Glass
Flasks, mixing cylinders and separatory funnels
Product Suggestions
Product Suggestions