Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
Supplier: DWK Life Sciences 8501000009
Description
- All stoppers are solid except Standard Taper stopper sizes 32 and 38, which are hollow
- Made in accordance with ASTM Standard E694
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
| Solid Pennyhead Stopper | |
| Glass | |
| Flasks, mixing cylinders and separatory funnels |