Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
Supplier:  DWK Life Sciences 8505002425
 View more versions of this product
Catalog No. K850500-2425
Only null left
Explore available promotions
Description
Description
  • Sizes Standard Taper 10/18 through Standard Taper 19/22 are solid
  • Sizes Standard Taper 24/25 and larger are hollow
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Solid Pennyhead Stopper
Glass
2.5 cm
24/25
Flasks, mixing cylinders and separatory funnels
24/25
Product Suggestions
Product Suggestions