Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
Supplier: DWK Life Sciences 8505001018
Description
- Sizes Standard Taper 10/18 through Standard Taper 19/22 are solid
- Sizes Standard Taper 24/25 and larger are hollow
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
| Solid Pennyhead Stopper | |
| Glass | |
| 1.8 cm |
| 10/18 | |
| Flasks, mixing cylinders and separatory funnels | |
| 10/18 |