Learn More
Thermo Scientific™ KingFisher™ Presto Purification System
Use the Thermo Scientific™ KingFisher™ Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
Spécifications
| Volume (Metric) Processing | 200 to 5000 μL |
|---|---|
| For Use With (Equipment) | KingFisher™ Presto Purification System |
| Samples per Run | 24 |
| Numéro de catalogue | Numéro du manufacturier. | Product Type | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Product Type | Prix | Quantity & Availability | |||||
501527929
|
Thermo Scientific™
24078841 |
24 DW head and heating block |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
501527928
|
Thermo Scientific™
24078840 |
24 Deep Well head |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Features:
- Purify 96 or 24 samples at a time
- Volume range of 50-5000μL
- Minimized size for easy fitting to automation systems
- Heating capability for application requiring elevated temperatures
- Easy connection to several liquid handling instruments
- Flexible system for wide variety of applications
Spécifications
| 200 to 5000 μL | |
| KingFisher™ Presto Purification System | |
| 24 |