Learn More
Thermo Scientific™ KingFisher™ Presto Purification System
Catalog No. 501527929
Encompass
Change view
Click to view available options
Product Type:
24 Deep Well head
24 DW head and heating block
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
501527929 24 DW head and heating block
501527928 24 Deep Well head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501527929 Supplier Thermo Scientific™ Supplier No. 24078841
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use the Thermo Scientific™ KingFisher™ Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
Features:
  • Purify 96 or 24 samples at a time
  • Volume range of 50-5000μL
  • Minimized size for easy fitting to automation systems
  • Heating capability for application requiring elevated temperatures
  • Easy connection to several liquid handling instruments
  • Flexible system for wide variety of applications
Specifications
Product Type 24 DW head and heating block
Volume (Metric) Processing 200 to 5000 μL
For Use With (Equipment) KingFisher™ Presto Purification System
Samples per Run 24