missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ Silicone Lids

Reusable lids stretch easily and adapt to many container sizes, improves lab safety, enhances sample management, offers a more sustainable and cost-effective solution over foils, films and other single-use covers.

Specifications

Material Silicone
For Use With (Application) To cover open lab vessels
For Use With (Equipment) Beakers, Bottles, Cylinders, Erlenmeyer Flask
Length (Metric) 16.00 cm
View More Specs

Products 12
Catalog Number Mfr. No. Color Description Price Quantity & Availability  
Catalog Number Mfr. No. Color Description Price Quantity & Availability  
15112171
View Documents Promotion Details
DWK Life Sciences
291111111
Pink Small N/A
Only null left
 
15112172
View Documents Promotion Details
DWK Life Sciences
291111128
Cyan Small N/A
Only null left
 
15112173
View Documents Promotion Details
DWK Life Sciences
291111136
Green Small N/A
Only null left
 
15112174
View Documents Promotion Details
DWK Life Sciences
291112116
Pink Medium N/A
Only null left
 
15112175
View Documents Promotion Details
DWK Life Sciences
291112124
Cyan Medium N/A
Only null left
 
15112176
View Documents Promotion Details
DWK Life Sciences
291112132
Green Medium N/A
Only null left
 
15112177
View Documents Promotion Details
DWK Life Sciences
291113112
Pink Large N/A
Only null left
 
15112178
View Documents Promotion Details
DWK Life Sciences
291113129
Cyan Large N/A
Only null left
 
15112179
View Documents Promotion Details
DWK Life Sciences
291113137
Green Large N/A
Only null left
 
15112180
View Documents Promotion Details
DWK Life Sciences
291110012
Pink Small, Medium & Large N/A
Only null left
 
15112181
View Documents Promotion Details
DWK Life Sciences
291110029
Cyan Small, Medium & Large N/A
Only null left
 
15112182
View Documents Promotion Details
DWK Life Sciences
291110037
Green Small, Medium & Large N/A
Only null left
 
Description

Description

VERSATILE

  • Fits on beakers, flasks, bottles, cylinders, plastic and glass products
  • Each size conforms to various diameters
  • Works with spouts, beads and thread finishes
  • Adaptable for many uses beyond limits of traditional films and covers

SAMPLE MANAGEMENT

  • Reduces cross-contamination of samples
  • Compatible with lab markers to enhance sample identification
  • Improves your lab workflow efficiency
  • Uses color to track projects and samples

LAB SAFETY

  • Improves safety performance over foils or films
  • Color aids quick visual identification
  • Protects skin and nose from contents
  • Defends against spill/leakage

SUSTAINABLE

  • Reusable, customizable and dishwasher safe
  • Reduces non-reusable consumables that generate waste such as films and foils
  • Chemically resistant, autoclavable and pliable over a wide temperature range
  • Reduces environmental impact and lowers lab operational costs
Specifications

Specifications

Silicone
To cover open lab vessels
Beakers, Bottles, Cylinders, Erlenmeyer Flask
16.00 cm
Videos
SDS
Documents

Documents