missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ Silicone Lids

Catalog No. 15112171
Change view
Click to view available options
Color:
Cyan
Green
Pink
Description:
Small, Medium & Large
Small
Medium
Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Description
15112171 Pink Small
15112172 Cyan Small
15112173 Green Small
15112174 Pink Medium
15112175 Cyan Medium
15112176 Green Medium
15112177 Pink Large
15112178 Cyan Large
15112179 Green Large
15112180 Pink Small, Medium & Large
15112181 Cyan Small, Medium & Large
15112182 Green Small, Medium & Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 missing translation for 'options'
Catalog No. 15112171 missing translation for 'mfr' DWK Life Sciences missing translation for 'supplierNo' 291111111

Reusable lids stretch easily and adapt to many container sizes, improves lab safety, enhances sample management, offers a more sustainable and cost-effective solution over foils, films and other single-use covers.

VERSATILE

  • Fits on beakers, flasks, bottles, cylinders, plastic and glass products
  • Each size conforms to various diameters
  • Works with spouts, beads and thread finishes
  • Adaptable for many uses beyond limits of traditional films and covers

SAMPLE MANAGEMENT

  • Reduces cross-contamination of samples
  • Compatible with lab markers to enhance sample identification
  • Improves your lab workflow efficiency
  • Uses color to track projects and samples

LAB SAFETY

  • Improves safety performance over foils or films
  • Color aids quick visual identification
  • Protects skin and nose from contents
  • Defends against spill/leakage

SUSTAINABLE

  • Reusable, customizable and dishwasher safe
  • Reduces non-reusable consumables that generate waste such as films and foils
  • Chemically resistant, autoclavable and pliable over a wide temperature range
  • Reduces environmental impact and lowers lab operational costs

Specifications

Color Pink
Description Small
Diameter (Metric) Outer 43 to 61 mm
Material Silicone
For Use With (Application) To cover open lab vessels
For Use With (Equipment) Beakers, Bottles, Cylinders, Erlenmeyer Flask
Height (Metric) 22.5 mm
Length (Metric) 16.00 cm
Quantity 1