Learn More
DWK Life Sciences KIMBLE™ Silicone Lids
Reusable lids stretch easily and adapt to many container sizes, improves lab safety, enhances sample management, offers a more sustainable and cost-effective solution over foils, films and other single-use covers.
Supplier:  DWK Life Sciences 291113112
 View more versions of this product
Catalog No. 15112177
Only null left
Description
Description
VERSATILE
  • Fits on beakers, flasks, bottles, cylinders, plastic and glass products
  • Each size conforms to various diameters
  • Works with spouts, beads and thread finishes
  • Adaptable for many uses beyond limits of traditional films and covers
SAMPLE MANAGEMENT
  • Reduces cross-contamination of samples
  • Compatible with lab markers to enhance sample identification
  • Improves your lab workflow efficiency
  • Uses color to track projects and samples
LAB SAFETY
  • Improves safety performance over foils or films
  • Color aids quick visual identification
  • Protects skin and nose from contents
  • Defends against spill/leakage
SUSTAINABLE
  • Reusable, customizable and dishwasher safe
  • Reduces non-reusable consumables that generate waste such as films and foils
  • Chemically resistant, autoclavable and pliable over a wide temperature range
  • Reduces environmental impact and lowers lab operational costs
Specifications
Chemical Identifiers
Silicone
To cover open lab vessels
Beakers, Bottles, Cylinders, Erlenmeyer Flask
16.00 cm
Specifications
Pink
84 to 116 mm
To cover open lab vessels
23.2 mm
1
Large
Silicone
Beakers, Bottles, Cylinders, Erlenmeyer Flask
16.00 cm
Product Suggestions
Product Suggestions