Learn More
DWK Life Sciences™ Kimble™ Polyacetal Spherical Joint Clamp
The Kimble™ Polyacetal Spherical Joint Clamp is an easy to use spherical joint positive closure clamp made of tough, color coded plastic for use with ball and socket joints.
$493.29 - $583.04
| Catalog Number | Mfr. No. | Standard Taper Joint | Color | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Standard Taper Joint | Color | Price | Quantity & Availability | ||||
|
01183038
|
DWK Life Sciences™
6753100013 |
12/5, 12/2, 12/3 | Violet |
Case of 12 Each for $493.29
|
|
||||
|
01183039
|
DWK Life Sciences™
6753100019 |
18/9, 18/7 | Light Blue |
Case of 12 Each for $583.04
|
|
||||
|
01183040
|
DWK Life Sciences™
6753100029 |
28/15, 28/12 | Dark Red |
Case of 12 Each for $553.89
|
|
||||
|
01183041
|
DWK Life Sciences™
6753100035 |
35/25, 35/20 | Light Orange |
Case of 12 Each for $563.50
|
|
||||
Description
- Manufactured from polyacetal
- This clamp has good chemical resistance at a lower cost than metal clips
- Clamp will not scratch glass and can be autoclaved to 200°C
- Easy, safe and quick to put on and take off
- Color coding per size
Specifications
| Polyacetal Spherical Joint Clamp | |
| Polyacetal |