Learn More
DWK Life Sciences™ Kimble™ Polyacetal Spherical Joint Clamp
The Kimble™ Polyacetal Spherical Joint Clamp is an easy to use spherical joint positive closure clamp made of tough, color coded plastic for use with ball and socket joints.
Supplier: DWK Life Sciences™ 6753100019
Description
- Manufactured from polyacetal
- This clamp has good chemical resistance at a lower cost than metal clips
- Clamp will not scratch glass and can be autoclaved to 200°C
- Easy, safe and quick to put on and take off
- Color coding per size
Chemical Identifiers
| Polyacetal Spherical Joint Clamp | |
| Polyacetal |
Specifications
| Polyacetal Spherical Joint Clamp | |
| Polyacetal | |
| 18/9, 18/7 | |
| Light Blue |