Learn More
DWK Life Sciences™ Kimble™ Polyacetal Spherical Joint Clamp
Description
- Manufactured from polyacetal
- This clamp has good chemical resistance at a lower cost than metal clips
- Clamp will not scratch glass and can be autoclaved to 200°C
- Easy, safe and quick to put on and take off
- Color coding per size
Specifications
Specifications
| Type | Polyacetal Spherical Joint Clamp |
| Material | Polyacetal |
| Standard Taper Joint | 28/15, 28/12 |
| Color | Dark Red |