missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Taperseal Liners (Cone Shaped)
Provides excellent sealing capabilities in liquid applications; seal is made across the top and the inside diameter of the container.
$227.82 - $6,256.50
Specifications
| Color | Black |
|---|---|
| Material | Phenolic Resin |
| Product Type | Phenolic Cap |
| For Use With (Equipment) | Bottle |
| Closure Liner | Taperseal |
| Catalog Number | Mfr. No. | Neck Finish | No. per Case | Packaging | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | No. per Case | Packaging | Price | Quantity & Availability | |||
|
13757093
|
DWK Life Sciences
7520520400 |
20-400 | 6300 | Bulk |
Case of 6300 Each for $3,926.85
|
|
|||
|
13757095
|
DWK Life Sciences
7520524400 |
24-400 | 4400 | Bulk |
Case of 4400 Each for $5,801.82
|
|
|||
|
13757097
|
DWK Life Sciences
7520533400 |
33-400 | 2400 | Bulk |
Case of 2300 Each for $4,005.43
|
|
|||
|
13757096
|
DWK Life Sciences
7520528400 |
28-400 | 3200 | Bulk |
Case of 3100 Each for $4,235.07
|
|
|||
|
13757094
|
DWK Life Sciences
7520522400 |
22-400 | 5300 | Bulk |
Case of 4700 Each for $6,256.50
|
|
|||
|
13757098
|
DWK Life Sciences
7520538400 |
38-400 | 1800 | Bulk |
Case of 1800 Each for $3,401.93
|
|
|||
|
13757169
|
DWK Life Sciences
75205G20400 |
20-400 | 144 | Case |
Case of 144 Each for $227.82
|
|
|||
|
13757170
|
DWK Life Sciences
75205G22400 |
22-400 | 144 | Case |
Case of 144 Each for $253.04
|
|
|||
|
13757171
|
DWK Life Sciences
75205G24400 |
24-400 | 144 | Case |
Case of 144 Each for $262.43
|
|
|||
|
13757172
|
DWK Life Sciences
75205G28400 |
28-400 | 144 | Case |
Case of 144 Each for $260.21
|
|
|||
|
13757173
|
DWK Life Sciences
75205G33400 |
33-400 | 144 | Case |
Case of 144 Each for $331.93
|
|
|||
|
13757174
|
DWK Life Sciences
75205G38400 |
38-400 | 144 | Case |
Case of 144 Each for $404.21
|
|
|||
Description
- Excellent sealing capability
- Commonly used for liquids
- Taperseal (cone shaped) LDPE liner provides a seal across the top and the inside diameter of the container
- Liner is stress crack resistant; exellent torque retention
All caps/liners should be tested to ensure leak and compatibility performance with contents.
Specifications
| Black | |
| Phenolic Cap | |
| Taperseal |
| Phenolic Resin | |
| Bottle |