Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Taperseal Liners (Cone Shaped)
Provides excellent sealing capabilities in liquid applications; seal is made across the top and the inside diameter of the container.
Supplier:  DWK Life Sciences 75205G33400
 Afficher plus de versions de ce produit
Numéro de catalogue. 13757173
Il en reste null
Description
Description
  • Phenolic screw cap with a cone-shaped taperseal low density polyethylene (LDPE) liner
  • Color: black
  • Bulk packed
  • Stress crack resistant
  • excellent torque retention
Spécifications
Identifiants chimiques
Black
Phenolic Cap
Taperseal
Phenolic Resin
Bottle
Spécifications
Black
144
33-400
Bottle
Phenolic Resin
Case
Phenolic Cap
Taperseal
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS