Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Taperseal Liners (Cone Shaped)
Provides excellent sealing capabilities in liquid applications; seal is made across the top and the inside diameter of the container.
Supplier: DWK Life Sciences 7520528400
Description
- Phenolic screw cap with a cone-shaped taperseal low density polyethylene (LDPE) liner
- Color: black
- Bulk packed
- Stress crack resistant
- excellent torque retention
Identifiants chimiques
| Black | |
| Phenolic Cap | |
| Taperseal |
| Phenolic Resin | |
| Bottle |
Spécifications
| Black | |
| 3200 | |
| 28-400 | |
| Bottle |
| Phenolic Resin | |
| Bulk | |
| Phenolic Cap | |
| Taperseal |