missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ Phenolic Caps with Taperseal Liners, Cone Shaped

Catalog No. 13757174
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
No. per Case:
144
1800
2400
3200
4400
5300
6300
Packaging:
Bulk
Case
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
13757174 38-400 144 Case
13757093 20-400 6300 Bulk
13757095 24-400 4400 Bulk
13757097 33-400 2400 Bulk
13757096 28-400 3200 Bulk
13757094 22-400 5300 Bulk
13757098 38-400 1800 Bulk
13757169 20-400 144 Case
13757170 22-400 144 Case
13757171 24-400 144 Case
13757172 28-400 144 Case
13757173 33-400 144 Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 13757174 Supplier DWK Life Sciences Supplier No. 75205G38400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ Phenolic Caps with Taperseal Liners (Cone Shaped) provide excellent sealing in liquid applications. The cone-shaped LDPE liner creates a two-part seal across the container rim and inner diameter, ensuring reliable closure with superior torque retention.

  • Phenolic screw cap with a cone-shaped taperseal low density polyethylene (LDPE) liner
  • Two-part seal: seals on the container rim and at the inner diameter of the container opening
  • Specially formulated phenolic resin resistant to stress cracking
  • Designed for superior torque retention
  • Excellent for sample storage and re-sealing
  • Color: Black
  • Bulk packed

Specifications

Color Black
Material Phenolic Resin
No. per Case 144
Packaging Case
Sterility Non-sterile
Neck Finish 38-400
For Use With (Equipment) Bottle
Closure Liner Taperseal
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Phenolic Cap
Show More Show Less