Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Pulp/Vinyl Liners
Cap provides good chemical resistance to mild acids, alkalis, alcohols, aqueous solutions, oils and solvents.
Supplier: DWK Life Sciences 75201G70400
Avertissement:
Not recommended for hydrocarbons or bleaches.
Description
- Polyvinyl film is adhered to 0.035-in. pulpboard
- Color: black
- Low cost cap for general purpose usage
- Not recommended for hydrocarbons or bleaches
Identifiants chimiques
| Black | |
| 144 Cs. | |
| Bottle |
| Phenolic Resin | |
| Phenolic Cap | |
| Pulp/Vinyl |
Spécifications
| Black | |
| 144 Cs. | |
| Phenolic Cap | |
| Pulp/Vinyl |
| Phenolic Resin | |
| 70-400 | |
| Bottle |