missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Pulp/Vinyl Liners
Cap provides good chemical resistance to mild acids, alkalis, alcohols, aqueous solutions, oils and solvents.
$95.23 - $696.29
Specifications
| Color | Black |
|---|---|
| Material | Phenolic Resin |
| No. per Case | 144 Cs. |
| Product Type | Phenolic Cap |
| For Use With (Equipment) | Bottle |
Alert:
Not recommended for hydrocarbons or bleaches.
| Catalog Number | Mfr. No. | Neck Finish | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | Price | Quantity & Availability | |||||
|
13757116
|
DWK Life Sciences
75201G28400 |
28-400 |
Case of 144 Each for $95.23
|
|
|||||
|
13757113
|
DWK Life Sciences
75201G20400 |
20-400 |
Case of 144 Each for $120.29
|
|
|||||
|
13757118
|
DWK Life Sciences
75201G38400 |
38-400 |
Case of 144 Each for $194.46
|
|
|||||
|
13757117
|
DWK Life Sciences
75201G33400 |
33-400 |
Case of 144 Each for $159.32
|
|
|||||
|
13757115
|
DWK Life Sciences
75201G24400 |
24-400 |
Case of 144 Each for $144.57
|
|
|||||
|
13757119
|
DWK Life Sciences
75201G43400 |
43-400 |
Case of 144 Each for $141.49
|
|
|||||
|
13757125
|
DWK Life Sciences
75201G70400 |
70-400 |
Case of 144 Each for $545.50
|
|
|||||
|
13757123
|
DWK Life Sciences
75201G58400 |
58-400 |
Case of 144 Each for $386.36
|
|
|||||
|
13757120
|
DWK Life Sciences
75201G45400 |
45-400 |
Case of 144 Each for $319.43
|
|
|||||
|
13757121
|
DWK Life Sciences
75201G48400 |
48-400 |
Case of 144 Each for $312.54
|
|
|||||
|
13757122
|
DWK Life Sciences
75201G53400 |
53-400 |
Case of 144 Each for $387.71
|
|
|||||
|
13757126
|
DWK Life Sciences
75201G89400 |
89-400 |
Case of 144 Each for $696.29
|
|
|||||
Description
- Low cost
- General purpose
- Temperature tolerant
- Good chemical resistance to mild acids, alkalis, alcohols, aqueous solutions, oils and solvents
All caps/liners should be tested to ensure leak and compatibility performance with contents.
Specifications
| Black | |
| 144 Cs. | |
| Bottle |
| Phenolic Resin | |
| Phenolic Cap | |
| Pulp/Vinyl |