missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences KIMBLE™ GLS 80™ KimCote™ Clear Wide Mouth Media Bottles with Caps
Get versatility, purity and durability to achieve results for laboratory storage, bioprocess and production applications.
$633.12 - $1,559.62
Specifications
| Bottle Surface | KimCote™ |
|---|---|
| Coating | KimCote™ |
| Color | Clear |
| For Use With (Application) | Storage and Transport |
| Material | Borosilicate Glass (3.3) |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
02913081
|
DWK Life Sciences
143900250 |
250 mL |
Case of 4 Each for $633.12
|
|
|||||
|
02913082
|
DWK Life Sciences
143900500 |
500 mL |
Case of 4 Each for $722.44
|
|
|||||
|
02913083
|
DWK Life Sciences
143901000 |
1000 mL |
Case of 4 Each for $903.26
|
|
|||||
|
02913084
|
DWK Life Sciences
143902000 |
2000 mL |
Case of 4 Each for $1,488.12
|
|
|||||
|
02913085
|
DWK Life Sciences
143905000 |
5000 mL |
Case for $1,559.62
|
|
|||||
Description
- Wide-mouth
- 3.3 borosilicate glass
- Durable ceramic ink graduations and marking patch
- Blue quick-release linerless polypropylene closure and pour ring are rated up to 140° C
- Autoclavable
- KimCote™ coating provides safety and reduces hazards of shattered glass
Specifications
| KimCote™ | |
| Clear | |
| Borosilicate Glass (3.3) | |
| Case | |
| Blue | |
| Yes | |
| Polypropylene | |
| Quick-release Screw Thread |
| KimCote™ | |
| Storage and Transport | |
| 140°C | |
| Non-sterile | |
| Yes | |
| Unlined | |
| GLS 80 | |
| Autoclavable |