missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ GLS 80™ KimCote™ Clear Wide Mouth Media Bottles with Caps

Get versatility, purity and durability to achieve results for laboratory storage, bioprocess and production applications.

Supplier:  DWK Life Sciences 143901000

 View more versions of this product

Catalog No. 02913083


Only null left

Description

Description

  • Wide-mouth
  • 3.3 borosilicate glass
  • Durable ceramic ink graduations and marking patch
  • Blue quick-release linerless polypropylene closure and pour ring are rated up to 140° C
  • Autoclavable
  • KimCote™ coating provides safety and reduces hazards of shattered glass
Specifications

Chemical Identifiers

KimCote™
Clear
Borosilicate Glass (3.3)
Case
Blue
Yes
Polypropylene
Quick-release Screw Thread
KimCote™
Storage and Transport
140°C
Non-sterile
Yes
Unlined
GLS 80
Autoclavable

Specifications

KimCote™
32 oz.
Clear
Blue Polypropylene Cap and Pouring Ring
225.425 mm
Case
Non-sterile
Yes
222 mm
Polypropylene
101 mm
Autoclavable
1000 mL
KimCote™
Storage and Transport
Borosilicate Glass (3.3)
140°C
4 Pk.
Blue
Yes
Unlined
GLS 80
Quick-release Screw Thread
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions