missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ GLS 80™ KimCote™ Clear Wide Mouth Media Bottles with Caps

Catalog No. 02913081
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
5000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02913081 250 mL
02913082 500 mL
02913083 1000 mL
02913084 2000 mL
02913085 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02913081 Supplier DWK Life Sciences Supplier No. 143900250

Get versatility, purity and durability to achieve results for laboratory storage, bioprocess and production applications.

  • Wide-mouth
  • 3.3 borosilicate glass
  • Durable ceramic ink graduations and marking patch
  • Blue quick-release linerless polypropylene closure and pour ring are rated up to 140° C
  • Autoclavable
  • KimCote™ coating provides safety and reduces hazards of shattered glass
Note: Image is representative only. Bottle capacity may vary.

Specifications

Bottle Surface KimCote™
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Coating KimCote™
Color Clear
For Use With (Application) Storage and Transport
Includes Blue Polypropylene Cap and Pouring Ring
Material Borosilicate Glass (3.3)
Length (Metric) 196.85 mm
Max. Temperature (Metric) 140°C
Packaging Case
Quantity 4 Pk.
Sterility Non-sterile
Closure Color Blue
Closure Included Yes
Graduated Yes
Height (Metric) 105 mm
Closure Liner Unlined
Closure Material Polypropylene
Closure Size GLS 80
Diameter (Metric) 95 mm
Closure Type Quick-release Screw Thread
Autoclavable Autoclavable
Show More Show Less