missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ GLS 80™ KimCote™ Clear Wide Mouth Media Bottles with Caps

Catalog No. 02913085
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
5000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02913085 5000 mL
02913081 250 mL
02913082 500 mL
02913083 1000 mL
02913084 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02913085 Supplier DWK Life Sciences Supplier No. 143905000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Get versatility, purity and durability to achieve results for laboratory storage, bioprocess and production applications.

  • Wide-mouth
  • 3.3 borosilicate glass
  • Durable ceramic ink graduations and marking patch
  • Blue quick-release linerless polypropylene closure and pour ring are rated up to 140° C
  • Autoclavable
  • KimCote™ coating provides safety and reduces hazards of shattered glass
Note: Image is representative only. Bottle capacity may vary.

Specifications

Bottle Surface KimCote™
Capacity (Metric) 5000 mL
Coating KimCote™
Color Clear
For Use With (Application) Storage and Transport
Includes Blue Polypropylene Cap
Material Borosilicate Glass (3.3)
Length (Metric) 196.85 mm
Max. Temperature (Metric) 140°C
Packaging Case
Quantity 1 Ea.
Sterility Non-sterile
Closure Color Blue
Closure Included Yes
Graduated Yes
Height (Metric) 314 mm
Closure Liner Unlined
Closure Material Polypropylene
Closure Size GLS 80
Diameter (Metric) 182 mm
Closure Type Quick-release Screw Thread
Autoclavable Autoclavable
Capacity (English) 170 oz.
Show More Show Less