Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached White Rubber Lined Caps
Packaged in corrugated trays with partitions
 Afficher plus de versions de ce produit
Numéro de catalogue. 14957211
Il en reste null
Description
Description
  • Small case quantities
  • Manufactured from clear 33 expansion borosilicate glass
  • Attached closed top black phenolic closures
Spécifications
Identifiants chimiques
A
Clear
Screw
Screw Cap (Closed-top)
Rubber
Borosilicate Glass
Phenolic
Spécifications
A
Clear
Borosilicate Glass
Screw
Phenolic
144/Cs.
Rubber
15 mm
4 mL
1 dr.
Screw Cap (Closed-top)
45 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS