missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached White Rubber Lined Caps

Packaged in corrugated trays with partitions

$332.21 - $694.03

Specifications

Classification A
Material (Liner) Rubber
Color Clear
Material Borosilicate Glass
Thread Style Screw
View More Specs

Products 12
Catalog Number Volume (Metric) Diameter (Metric) Outer Height (Metric) Quantity Price Quantity & Availability  
Catalog Number Volume (Metric) Diameter (Metric) Outer Height (Metric) Quantity Price Quantity & Availability  
14957222
View Documents Promotion Details
40 mL 28 mm 95 mm 72/Cs.
Case of 72 Each for $332.21
Only null left
 
14957220
View Documents Promotion Details
24 mL 23 mm 85 mm 144/Cs.
Case of 144 Each for $672.14
Only null left
 
14957221
View Documents Promotion Details
25 mL 28 mm 70 mm 72/Cs.
Case of 72 Each for $334.52
Only null left
 
14957214
View Documents Promotion Details
4 mL 17 mm 38 mm 200/Cs.
Case of 200 Each for $612.21
Only null left
 
14957216
View Documents Promotion Details
8 mL 17 mm 60 mm 144/Cs.
Case of 144 Each for $428.66
Only null left
 
14957218
View Documents Promotion Details
16 mL 21 mm 70 mm 144/Cs.
Case of 144 Each for $503.97
Only null left
 
14957212
View Documents Promotion Details
2 mL 12 mm 35 mm 288/Cs.
Case of 288 Each for $694.03
Only null left
 
14957213
View Documents Promotion Details
2 mL 15 mm 28 mm 200/Cs.
Case of 200 Each for $586.41
Only null left
 
14957215
View Documents Promotion Details
6 mL 19 mm 40 mm 200/Cs.
Case of 200 Each for $561.21
Only null left
 
14957219
View Documents Promotion Details
20 mL 28 mm 57 mm 72/Cs.
Case of 72 Each for $352.45
Only null left
 
14957217
View Documents Promotion Details
12 mL 19 mm 65 mm 144/Cs.
Case of 144 Each for $379.45
Only null left
 
14957211
View Documents Promotion Details
4 mL 15 mm 45 mm 144/Cs.
Case of 144 Each for $428.66
Only null left
 
Description

Description

  • Small case quantities
  • Manufactured from clear 33 expansion borosilicate glass
  • Attached closed top black phenolic closures

Specifications

Specifications

A
Clear
Screw
Screw Cap (Closed-top)
Rubber
Borosilicate Glass
Phenolic
Videos
SDS
Documents

Documents