Learn More
DWK Life Sciences Kimble™ KIMAX™ Accuform™ Micro-Vial™ Vials
Conical cavity allows maximum retrieval of contents with a syringe needle.
Supplier:  DWK Life Sciences 6071010
 Afficher plus de versions de ce produit
Numéro de catalogue. 06100Q
Il en reste null
Description
Description
  • Clear glass Micro-Vial with open top closure and PTFE-faced silicone septum
  • Made with extra thick clear glass walls for both sturdiness and magnification of contents
  • Ungraduated, with marking spot
  • Tube has a screw thread finish
Spécifications
Identifiants chimiques
PTFE/Silicone
Tapered
Screw Cap (Open-top)
Borosilicate Glass
Screw
Spécifications
PTFE/Silicone
25 mm
10.0 mL
Tapered
2.75 dr.
ASTM E438, USP Type I, Type I, Class A
Clear
Borosilicate Glass
No
Screw
Screw Cap (Open-top)
73 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS