Learn More
DWK Life Sciences Kimble™ KIMAX™ Accuform™ Micro-Vial™ Vials
Conical cavity allows maximum retrieval of contents with a syringe needle.
$521.14 - $921.11
Specifications
Material (Liner) PTFE/Silicone
Material Borosilicate Glass
Style Tapered
Thread Style Screw
Closure Type Screw Cap (Open-top)
View More Specs
Products 7
Catalog Number Mfr. No. Volume (Metric) Color Graduated Volume (English) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Color Graduated Volume (English) Price Quantity & Availability  
06100A
View Documents Promotion Details
DWK Life Sciences
60710110
0.1 mL Clear Yes 0.03 dr.
Case of 12 Each for $521.14
Only null left
06100B
View Documents Promotion Details
DWK Life Sciences
60710310
0.3 mL Clear Yes 0.08 dr.
Case of 12 Each for $636.14
Only null left
06100H
View Documents Promotion Details
DWK Life Sciences
607101
1.0 mL Clear No 0.25 dr.
Case of 12 Each for $669.50
Only null left
06100J
View Documents Promotion Details
DWK Life Sciences
607102
2.0 mL Clear No 0.5 dr.
Case of 12 Each for $707.61
Only null left
06100K
View Documents Promotion Details
DWK Life Sciences
607105
5.0 mL Clear No 1.35 dr.
Case of 12 Each for $811.14
Only null left
06100Q
View Documents Promotion Details
DWK Life Sciences
6071010
10.0 mL Clear No 2.75 dr.
Case of 12 Each for $921.11
Only null left
06100R
View Documents Promotion Details
DWK Life Sciences
607103
3.0 mL Clear No 0.8 dr.
Case of 12 Each for $727.32
Only null left
Description
Description
  • Clear glass Micro-Vial with open top closure and PTFE-faced silicone septum
  • Made with extra thick clear glass walls for both sturdiness and magnification of contents
  • Ungraduated, with marking spot
  • Tube has a screw thread finish
Specifications
Specifications
PTFE/Silicone
Tapered
Screw Cap (Open-top)
Borosilicate Glass
Screw