missing translation for 'onlineSavingsMsg'
Learn More

Ansell KleenGuard™ A20 Breathable Particle Protection Shoe Covers
GREENER_CHOICE

Catalog No. 19005059
Change view
Click to view available options
Closure Type:
Elastic Ankle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
19005059 Elastic Ankle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19005059 Supplier Ansell Supplier No. 66857

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seamless design

  • Packaged for cleanrooms (double bagged) and feature seamless elastic sole, toe and heel
  • Popular choice for laboratories and remediation, as well as pesticide applications
  • Air and moisture vapor can pass through to aid wearer comfort
  • Triple-stitched serged seams
  • Can withstand multiple wearing
  • Made of durable, breathable patented Microforce™ barrier SMS fabric
  • Provide particle control and comfort, as well as traction for slip resistance
  • Color: Light Blue

Specifications

Size X-Large/2X-Large
Material SMS
Color Light Blue
Closure Type Elastic Ankle
Sole Type Non-skid
Seam Type 3-stitched serged
For Use With (Application) Ideal for cleanroom use and for laboratories, decontamination, remediation, pesticides, heavy dust environments
Certifications/Compliance NFPA 99
Product Type Controlled Environments Shoe Covers