missing translation for 'onlineSavingsMsg'
Learn More

Kimtech™ M6 Pleated-Style Cleanroom Face Mask
GREENER_CHOICE

Catalog No. 189994781
Change view
Click to view available options
Style:
Pleated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
189994781 Pleated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 189994781 Supplier Kimtech™ Supplier No. 62477
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recommended for ISO Class 6 or higher cleanrooms

  • For general-use in low-end cleanrooms
  • Two-layer design with knitted earloops
  • Apertured low-linting outer fabric for protection and durability
  • BiCoSof™ fabric inner facing provides comfort
  • Double-bagged

Specifications

Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material Polyethylene
Size Standard
Sterility Non-sterile
Style Pleated
Cleanroom Class Class 1000 (ISO 6)
Construction 2-ply
Disposable Single-use
Filter Efficiency 89% (PFE), 39.3% (BFE)
For Use With (Application) Semiconductors, Disk Drives, Data Storage, Integrated Circuits
Latex Free Yes
Model 62477
Pore Size 0.1 μm (PFE), 3 μm (BFE)
Series Kimtech™ M6
Show More Show Less