missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech Pure™ A5 Cleanroom Non-Sterile Hoods
GREENER_CHOICE

Catalog No. 19812863
Change view
Click to view available options
Closure Type:
None
Ties
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
19812863 None
19169868 Ties
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19812863 Supplier Ansell Supplier No. 88849
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Non-sterile cleanroom hoods cover the head, neck, and shoulders to prevent contamination in the environment. Availble with or without ties.

  • Non-sterile, white
  • Triple bagged, vacuum packed
  • Suitable for ISO Class 5 or higher cleanrooms; CE certified
  • Works with a variety of masks
  • Latex-free
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions

Specifications

Product Type Open Face
Size Universal
Material Polypropylene SMS Fabric
Color White
Closure Type None
Latex Free Yes
Model 88849
Sterility Non-sterile
Packaging Type Triple-bagged, Vacuum Packaged
Quantity 100/Cs.
Includes No ties
For Use With (Application) ISO 5 to 8 cleanroom environments, compounding, mixing, filling, cleaning
Cleanroom Class Class 100 (ISO 5)
Show More Show Less