Learn More
KleenGuard™ Unispec™ II Safety Glasses
GREENER_CHOICE
Catalog No. 19041232
Change view
Click to view available options
Lens Tint:
Clear
Catalog No. Lens Tint
19041232 Clear
1 options
Catalog No. 19041232 Supplier KleenGuard™ Supplier No. 25646
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KleenGuard™ Unispec II Safety Eyewear offers wraparound protection to fit over most prescription eyewear.
  • Low-cost wraparound protection with one-piece clear polycarbonate lens
  • Uncoated clear frame
  • Side shields provide added protection
Compliance
Meets ANSI Z87.1+ 2010 standards
Specifications
Certifications/Compliance CE Approved, ANSI Z87.1/CSA Z94.3
Lens Coating Uncoated
Lens Tint Clear
Product Type Safety Glasses
Material Plastic
Disposable Reusable
Lens Material Plastic
Frame Material Plastic
UV Protection UV400
Frame Type Frameless