Learn More
DWK Life Sciences Kimble™ KIMAX™ Weighing Bottles — With standard taper Outside Ground Caps
Tall, cylindrical form with standard taper caps
$523.91 - $1,844.79
Specifications
Material Glass
Shape Cylindrical
Closure Material Glass
Closure Type Cap
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity & Availability  
03422C
View Documents Promotion Details
DWK Life Sciences
151462540
12 mL 29/12
Pack of 6 Each for $523.91
Case of 3 Pack for $1,571.73
Only null left
03422D
View Documents Promotion Details
DWK Life Sciences
151462550
16 mL 29/12
Case of 18 Each for $1,571.74
Only null left
03422E
View Documents Promotion Details
DWK Life Sciences
151463060
30 mL 34/12
Pack of 6 Each for $653.61
Case of 3 Pack for $1,960.83
Only null left
03422F
View Documents Promotion Details
DWK Life Sciences
151464050
45 mL 45/12
Case of 12 Each for $1,612.53
Only null left
03422G
View Documents Promotion Details
DWK Life Sciences
151464080
70 mL 45/12
Pack of 6 Each for $920.32
Case of 2 Pack for $1,840.64
Only null left
03422H
View Documents Promotion Details
DWK Life Sciences
1514640100
92 mL 45/12
Case of 12 Each for $1,844.79
Only null left
Description
Description
Tall, cylindrical form with standard taper caps
With Outside standard taper Ground Caps
  • Caps protect bottle in case of tipping; have open bottom
  • Prevent material from touching cap edges, provide increased headroom
  • Designed from Fed. Spec. NNN-B-1493, Type I, Style II
Specifications
Specifications
Glass
Cylindrical
Glass
Cap