missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Polypropylene Hoods

Catalog No. 19068502
Change view
Click to view available options
Product Type:
Hood, Crown Style
Hood, Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19068502 Hood, Crown Style
19068501 Hood, Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19068502 Supplier Keystone Safety Supplier No. HNWIECROWN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

One-size-fits-all, latex-free, breathable hoods provide protection from contamination.

100% spunbond polypropylene defends against contamination and is comfortable to wear

 

  • Lightweight and disposable
  • Latex-free elastic closure
  • Flame-resistant
  • Universal size
  • Color: White
The crown style has stitching on the top for a better fit, and offers an overall more spacious wider design in situations where more room is required.

 

Specifications

Product Type Hood, Crown Style
Size Universal
Material Polypropylene
Color White
Closure Type Elastic
Disposable Single-use
Seam Type Serged
Packaging Type 100/bag (10 bags/case)
Quantity 1000/Cs.
Length (English) 14 in.
Length (Metric) 35.56 cm
For Use With (Application) Dry Particulates, Dirt, and Grime
Show More Show Less