Learn More
Keystone Safety Heavy Duty Polypropylene Cleanroom Lab Coats
Lightweight, latex-free lab coats are made from high quality materials and compatible in Class ISO7 cleanroom environments.
Supplier:  Keystone Safety LC0WENWHDSMWHITE
 View more versions of this product
Catalog No. 19-068-530
Only null left
Explore available promotions
Description
Description
Protect workers against dry particulates, dirt and grime
  • Non-woven, nonlinting, spunbond heavyweight polypropylene construction
  • Serged seams offer high tensile strength
  • Latex-free
  • Disposable
  • Elastic wrists, snap front, single collar, no pockets
  • Available in six sizes
  • Flame resistant
Ideal for Class 10,000 environments and above. Commonly used in electronic, medical device and pharmaceutical industries where operator garment control is a concern.
Specifications
Specifications
Controlled Environments Lab Coat
Serged
Class 10,000 (ISO 7)
Single-use
Electronics, Medical Device, Pharmaceutical
Product Suggestions
Product Suggestions