missing translation for 'onlineSavingsMsg'
Learn More

Josamycin, 98%

CAS: 16846-24-5 | C42H69NO15 | 828.01 g/mol

Supplier:  Thermo Scientific Chemicals J62245MF

Catalog No. AAJ62245MF

Only null left
Explore available promotions

Description

Description

Josamycin, is used as a macrolide antibiotic effective against a variety of pathogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Josamycin, is used as a macrolide antibiotic effective against a variety of pathogens.

Solubility
Soluble in ethanol

Notes
Protect from light and moisture. Store in cool place. Keep container tightl
Specifications

Specifications

Josamycin
Powder
White
C42H69NO15
14,5267
Soluble in ethanol
CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]1O[C@H](C)[C@@H](O[C@H]2C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O2)[C@@H]([C@H]1O)N(C)C)OC(C)=O
828.01
827.99
Josamycin
16846-24-5
50 mg
MFCD00210320
Leucomycin A3
XJSFLOJWULLJQS-NGVXBBESSA-N
(2S,3S,4R,6S)-6-{[(2R,3S,4R,5R,6S)-6-{[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-(acetyloxy)-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-6-yl]oxy}-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy}-4-hydroxy-2,4-dimethyloxan-3-yl 3-methylbutanoate
131849444
≥98%

Chemical Identifiers

16846-24-5
828.01
XJSFLOJWULLJQS-NGVXBBESSA-N
131849444
CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]1O[C@H](C)[C@@H](O[C@H]2C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O2)[C@@H]([C@H]1O)N(C)C)OC(C)=O
C42H69NO15
MFCD00210320
Leucomycin A3
(2S,3S,4R,6S)-6-{[(2R,3S,4R,5R,6S)-6-{[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-(acetyloxy)-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-6-yl]oxy}-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy}-4-hydroxy-2,4-dimethyloxan-3-yl 3-methylbutanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 240-871-6

RTECSNumber : OH4725810

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only