missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Standard Single Element Ion Standard - Cation, SPEX CertiPrep™
Description
Comprend
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| NH4+ | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
Spécifications
| Packaging | Triple-leached LDPE bottle |
| Certificate of Analysis | With Certificate of Analysis |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| CAS Min % | 0.1, 99.9 |
| CAS Max % | 0.1, 99.9 |
| CAS | 7732-18-5 |
| Technique | Ion Chromatography |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Solution Type | Standard Solution |
| Type | Certified Reference Material |
| Afficher plus de résultats |
Sécurité et manipulation
| DOTInformation | Nonhazardous |
| Recommended Storage | Ambient |
Product intended for laboratory use only